Learn More
New England Biolabs, Inc. XmnI – 5000 units
SDP
Catalog No. 50812122
Encompass
A restriction endonuclease that recognizes the sequence GAANN_^NNTTC.
Catalog No. 50-812-122 Supplier New England Biolabs, Inc. Supplier No. R0194L
A restriction endonuclease that recognizes the sequence GAANN_^NNTTC.