Learn More
TARGETMOL CHEMICALS INC XMU-MP-1 10MG
SDP
Catalog No. 502579927
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. XMU-MP-1 is an inhibitor of the pro-apoptotic sterile 20-like kinases MST1 and 2. purity: 99%
Catalog No. 50-257-9927 Supplier TARGETMOL CHEMICALS INC Supplier No. T421210MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. XMU-MP-1 is an inhibitor of the pro-apoptotic sterile 20-like kinases MST1 and 2. purity: 99%