Learn More
Medchemexpress LLC HY-100526 2mg Medchemexpress, XMU-MP-1 CAS:2061980-01-4 Purity:>98%
SDP
Catalog No. 502028627
Encompass_Preferred
Medchemexpress, HY-100526 2mg XMU-MP-1 CAS:2061980-01-4 XMU-MP-1 is a reversible and selective MST1/2 inhibitor with IC50s of 71.1 and 38.1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8627 Supplier Medchemexpress LLC Supplier No. HY1005262MG
Medchemexpress, HY-100526 2mg XMU-MP-1 CAS:2061980-01-4 XMU-MP-1 is a reversible and selective MST1/2 inhibitor with IC50s of 71.1 and 38.1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.