Learn More
Selleck Chemical LLC XMU-MP-1 2mg 2061980-01-4
SDP
Supplier:  Selleck Chemical LLC S83342MG
Encompass_Preferred
XMU-MP-1 is an inhibitor of MST1/2 with IC50 values of 71.1¿12.9 nM and 38.1¿6.9 nM against MST1 and MST2, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5960