Learn More
Medchemexpress LLC Xmu-mp-3 | 2031152-08-4 | 95.7% | C27H27F3N8O | 10MG
SDP
Catalog No. 5000261212
Encompass_Preferred
XMU-MP-3 is a potent, non-covalent inhibitor of Bruton's tyrosine kinase (BTK) that inhibits both wild-type BTK and the C481S mutant. It demonstrates low-nanomolar biochemical potency and cellular activity, including induction of apoptosis, and is suitable for biochemical and cellular studies of BTK signaling and inhibitor profiling.
  • Low-nanomolar biochemical potency (IC50 10.7 nM for BTK WT, 17.0 nM for BTK C481S in the presence of 10 μM ATP).
  • Demonstrated cellular activity (BTK-transformed Ba/F3 proliferation IC50 11.4 nM).
  • Induces apoptosis in BTK-dependent cell models.
  • High chemical purity (95.7%).
  • Defined storage stability for powder and solutions.
  • Suitable for biochemical assays and cellular mechanistic studies.
Catalog No. 50-002-61212 Supplier Medchemexpress LLC Supplier No. HY13653110MG
XMU-MP-3 is a potent, non-covalent inhibitor of Bruton's tyrosine kinase (BTK) that inhibits both wild-type BTK and the C481S mutant. It demonstrates low-nanomolar biochemical potency and cellular activity, including induction of apoptosis, and is suitable for biochemical and cellular studies of BTK signaling and inhibitor profiling.
  • Low-nanomolar biochemical potency (IC50 10.7 nM for BTK WT, 17.0 nM for BTK C481S in the presence of 10 μM ATP).
  • Demonstrated cellular activity (BTK-transformed Ba/F3 proliferation IC50 11.4 nM).
  • Induces apoptosis in BTK-dependent cell models.
  • High chemical purity (95.7%).
  • Defined storage stability for powder and solutions.
  • Suitable for biochemical assays and cellular mechanistic studies.