Learn More
Medchemexpress LLC XMU-MP-3 | 2031152-08-4 | 95.78% | 536.55 | 100 MG
SDP
Catalog No. 5000348709
Encompass_Preferred
XMU-MP-3 is a potent non-covalent BTK inhibitor with IC50s of 10.7 nM for BTK WT and 17.0 nM for BTK C481S mutation in the presence of 10 μM ATP. It also induces apoptosis. This product is for research use only.
  • Potent non-covalent BTK inhibitor
  • Induces apoptosis
  • Inhibits BTK-transformed Ba/F3 cell proliferation
  • Maintains inhibitory potency against BTK(C481S)-Ba/F3 cells
  • Substantially suppresses tumor growth in mouse xenograft models
  • Significantly reduces tumor size without affecting animal weights
Catalog No. 50-003-48709 Supplier Medchemexpress LLC Supplier No. HY136531100MG
XMU-MP-3 is a potent non-covalent BTK inhibitor with IC50s of 10.7 nM for BTK WT and 17.0 nM for BTK C481S mutation in the presence of 10 μM ATP. It also induces apoptosis. This product is for research use only.
  • Potent non-covalent BTK inhibitor
  • Induces apoptosis
  • Inhibits BTK-transformed Ba/F3 cell proliferation
  • Maintains inhibitory potency against BTK(C481S)-Ba/F3 cells
  • Substantially suppresses tumor growth in mouse xenograft models
  • Significantly reduces tumor size without affecting animal weights