Learn More
Medchemexpress LLC XMU-MP-3 | 2031152-08-4 | MFCD34470527; MFCD19703308 | 95.7% | 536.55 g·mol⁻¹ | C27H27F3N8O | 5 MG
SDP
Catalog No. 5000222522
Encompass_Preferred
XMU-MP-3 is a non-covalent small-molecule inhibitor of Bruton's tyrosine kinase (BTK) with low-nanomolar biochemical potency against wild-type and C481S mutant BTK. It shows activity in BTK-dependent cellular proliferation assays and is supplied as a solid research compound with reported purity around 95.7%.
  • Potent non-covalent inhibition of BTK (WT and C481S)
  • Low-nanomolar biochemical potency suitable for in vitro studies
  • Demonstrated cellular activity in BTK-dependent proliferation assays
  • Soluble in DMSO for assay preparation
  • Available in small solid packs for preclinical research
Catalog No. 50-002-22522 Supplier Medchemexpress LLC Supplier No. HY1365315MG
XMU-MP-3 is a non-covalent small-molecule inhibitor of Bruton's tyrosine kinase (BTK) with low-nanomolar biochemical potency against wild-type and C481S mutant BTK. It shows activity in BTK-dependent cellular proliferation assays and is supplied as a solid research compound with reported purity around 95.7%.
  • Potent non-covalent inhibition of BTK (WT and C481S)
  • Low-nanomolar biochemical potency suitable for in vitro studies
  • Demonstrated cellular activity in BTK-dependent proliferation assays
  • Soluble in DMSO for assay preparation
  • Available in small solid packs for preclinical research