Learn More
Cayman Chemical Xanthine/Hypoxanthine Fluorometric/Colorimetric Assay Kit, 96 Well
SDP
Catalog No. 502892438
Encompass_Preferred
Cayman’s Xanthine/Hypoxanthine Assay Kit provides fluorometric and colorimetric methods for measuring xanthine and hypoxanthine in tissue homogenates, cell lysates, cell culture supernatants, and biological fluids, such as serum, plasma, and urine.; In the assay, XO catalyzes the conversion of xanthine and hypoxanthine to uric acid and hydrogen peroxide (H2O2).; The fluorescence of this compound can be quantified at excitation and emission wavelengths of 535 and 587 nm, respectively.; Alternatively, the absorbance can be measured at 570 nm.; Background signal generated from H2O2 and other interferences found inherently in samples that are not produced by XO in the assay can be corrected by omitting the enzyme in sample background reactions.; When read fluorometrically, this assay has a range of 0.04-5 µM and a limit of detection (LOD) of 0.04 µM.; When read colorimetrically, the range is 0.18-100 µM with a LOD of 0.18 µM.
Catalog No. 50-289-2438 Supplier Cayman Chemical Supplier No. 70234096 WELL
Cayman’s Xanthine/Hypoxanthine Assay Kit provides fluorometric and colorimetric methods for measuring xanthine and hypoxanthine in tissue homogenates, cell lysates, cell culture supernatants, and biological fluids, such as serum, plasma, and urine.; In the assay, XO catalyzes the conversion of xanthine and hypoxanthine to uric acid and hydrogen peroxide (H2O2).; The fluorescence of this compound can be quantified at excitation and emission wavelengths of 535 and 587 nm, respectively.; Alternatively, the absorbance can be measured at 570 nm.; Background signal generated from H2O2 and other interferences found inherently in samples that are not produced by XO in the assay can be corrected by omitting the enzyme in sample background reactions.; When read fluorometrically, this assay has a range of 0.04-5 µM and a limit of detection (LOD) of 0.04 µM.; When read colorimetrically, the range is 0.18-100 µM with a LOD of 0.18 µM.