Learn More
Cayman Chemical Xanthohumol, Purity: ≥98%, CAS: 6754-58-1, 10 mg
SDP
Catalog No. 502971683
Encompass_Preferred
A natural prenylated chalcone that induces protective detoxification enzymes, at least in part via the Nrf2 pathway; inhibits PDK1 (IC50 = 6.6 μM) but not PKC in an in vitro assay; also can have anti-inflammatory, antioxidant, anti-carcinogenic, and osteogenic effects
Catalog No. 50-297-1683 Supplier Cayman Chemical Supplier No. 1539910 MG
A natural prenylated chalcone that induces protective detoxification enzymes, at least in part via the Nrf2 pathway; inhibits PDK1 (IC50 = 6.6 μM) but not PKC in an in vitro assay; also can have anti-inflammatory, antioxidant, anti-carcinogenic, and osteogenic effects