Learn More
Cayman Chemical Xanthosine 5'-monophosphate (sodium salt), Purity: ≥95%, CAS: 25899-70-1, 100 mg
SDP
Catalog No. 502968491
Encompass_Preferred
An intermediate in purine metabolism, the product of the rate limiting step in the generation of guanosine monophosphate, which is important for DNA, RNA, and glycoprotein synthesis.
Catalog No. 50-296-8491 Supplier Cayman Chemical Supplier No. 18134100 MG
An intermediate in purine metabolism, the product of the rate limiting step in the generation of guanosine monophosphate, which is important for DNA, RNA, and glycoprotein synthesis.