Learn More
Cayman Chemical XnthosIn 5-monphosphate 10mg
SDP
Catalog No. 502964898
Encompass_Preferred
An intermediate in purine metabolism, the product of the rate limiting step in the generation of guanosine monophosphate, which is important for DNA, RNA, and glycoprotein synthesis.
Catalog No. 50-296-4898 Supplier Cayman Chemical Supplier No. 1813410 MG
An intermediate in purine metabolism, the product of the rate limiting step in the generation of guanosine monophosphate, which is important for DNA, RNA, and glycoprotein synthesis.