Learn More
Cayman Chemical XnthosIn 5-monphosphate 50mg
SDP
Catalog No. 502966684
Encompass_Preferred
An intermediate in purine metabolism, the product of the rate limiting step in the generation of guanosine monophosphate, which is important for DNA, RNA, and glycoprotein synthesis.
Catalog No. 50-296-6684 Supplier Cayman Chemical Supplier No. 1813450 MG
An intermediate in purine metabolism, the product of the rate limiting step in the generation of guanosine monophosphate, which is important for DNA, RNA, and glycoprotein synthesis.