Learn More
Cayman Chemical Xanthurenic Acid, Purity: ≥95%, CAS: 59-00-7, 500 mg
SDP
Catalog No. 502981637
Encompass_Preferred
An endogenous mGluR2 and mGluR3 agonist implicated in the pathophysiology of schizophrenia; induces gametogenesis of P. falciparum
Catalog No. 50-298-1637 Supplier Cayman Chemical Supplier No. 19191500 MG
An endogenous mGluR2 and mGluR3 agonist implicated in the pathophysiology of schizophrenia; induces gametogenesis of P. falciparum