Learn More
Medchemexpress LLC XP-524 | 2344825-52-9 | 98.0% | 552.65 | 100 MG
SDP
Catalog No. 5000359885
Encompass_Preferred
XP-524 is a potent BET and EP300 inhibitor that demonstrates significant tumoricidal activity in vivo. It prevents KRAS-induced neoplastic transformation and extends survival in aggressive pancreatic ductal adenocarcinoma (PDAC) mouse models. This compound also enhances self-peptide presentation and tumor recruitment of cytotoxic T lymphocytes, highlighting its potential for PDAC research.
  • Potent BET and EP300 inhibitor
  • Exhibits significant tumoricidal activity in vivo
  • Prevents KRAS-induced neoplastic transformation
  • Extends survival in aggressive pancreatic ductal adenocarcinoma (PDAC) mouse models
  • Enhances self-peptide presentation and tumor recruitment of cytotoxic T lymphocytes
  • Effective against fulvestrant-resistant and tamoxifen-resistant/sensitive MCF7/T47D cells
  • Stable as powder for 3 years at -20°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C
Catalog No. 50-003-59885 Supplier Medchemexpress LLC Supplier No. HY147008100MG
XP-524 is a potent BET and EP300 inhibitor that demonstrates significant tumoricidal activity in vivo. It prevents KRAS-induced neoplastic transformation and extends survival in aggressive pancreatic ductal adenocarcinoma (PDAC) mouse models. This compound also enhances self-peptide presentation and tumor recruitment of cytotoxic T lymphocytes, highlighting its potential for PDAC research.
  • Potent BET and EP300 inhibitor
  • Exhibits significant tumoricidal activity in vivo
  • Prevents KRAS-induced neoplastic transformation
  • Extends survival in aggressive pancreatic ductal adenocarcinoma (PDAC) mouse models
  • Enhances self-peptide presentation and tumor recruitment of cytotoxic T lymphocytes
  • Effective against fulvestrant-resistant and tamoxifen-resistant/sensitive MCF7/T47D cells
  • Stable as powder for 3 years at -20°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C