Learn More
System Biosciences LLC XPack-GFP-loaded HEK293 Exosomes = 50ug
SDP
Catalog No. NC1785508
Encompass_Preferred
Deliver GFP to cells or simply track GFP-labeled exosomes with our ready-to-use XPack-GFP-loaded HEK293 Exosomes. Want an easy way to label cells with GFP? Looking for already GFP-labeled exosomes? SBI’s got you covered with our XPack-GFP-loaded HEK293 Exosomes. These isolated exosomes come pre-loaded with GFP and are ready for use in your experiments.
Catalog No. NC1785508 Supplier System Biosciences LLC Supplier No. XPAK100EXG
Deliver GFP to cells or simply track GFP-labeled exosomes with our ready-to-use XPack-GFP-loaded HEK293 Exosomes. Want an easy way to label cells with GFP? Looking for already GFP-labeled exosomes? SBI’s got you covered with our XPack-GFP-loaded HEK293 Exosomes. These isolated exosomes come pre-loaded with GFP and are ready for use in your experiments.