Learn More
Medchemexpress LLC XPC-6444 | 2230144-21-3 | 98.9% | 50 MG
SDP
Catalog No. 5000338354
Encompass_Preferred
XPC-6444 is a highly potent, isoform-selective, and CNS-penetrant inhibitor of NaV1.6, with an IC50 of 41 nM for hNaV1.6. It also effectively blocks NaV1.2, showing an IC50 of 125 nM. This compound exhibits anticonvulsant activity.
  • Highly potent and isoform-selective NaV1.6 inhibitor.
  • Effective in blocking NaV1.2.
  • Demonstrates anticonvulsant activity.
  • Shows good metabolic stability in human liver microsomes and hepatocytes.
  • Possesses low potential for MDR1 mediated efflux.
  • CNS-penetrant.
Catalog No. 50-003-38354 Supplier Medchemexpress LLC Supplier No. HY12877250MG
XPC-6444 is a highly potent, isoform-selective, and CNS-penetrant inhibitor of NaV1.6, with an IC50 of 41 nM for hNaV1.6. It also effectively blocks NaV1.2, showing an IC50 of 125 nM. This compound exhibits anticonvulsant activity.
  • Highly potent and isoform-selective NaV1.6 inhibitor.
  • Effective in blocking NaV1.2.
  • Demonstrates anticonvulsant activity.
  • Shows good metabolic stability in human liver microsomes and hepatocytes.
  • Possesses low potential for MDR1 mediated efflux.
  • CNS-penetrant.