Learn More
Thermo Scientific™ Xpect™ C. difficile Toxin A/B Test
GSA_VA
Catalog No. R24650 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 Tests/Kit
40 Tests/Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R24650 20 Tests/Kit
R24640 40 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R24650 Supplier Thermo Scientific™ Supplier No. R24650
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rapidly detect Clostridium difficile toxin A and B from human fecal samples with a simple, three-step procedure and results in 20 minutes.
Rapidly detect Clostridium difficile toxin A and B from human fecal specimens using Thermo Scientific™ Xpect™ C. difficile Toxin A/B Test. Combine excellent clinical performance with a simple, three-step procedure that offers accurate results within 20 minutes with less than one minute hands-on time. When you need STAT testing or 24/7 coverage, choose Xpect.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Clostridium difficile toxin A and B. Enhance laboratory performance and efficiency with excellent clinical sensitivity and specificity and a simple walk-away procedure.
Xpect C. difficile Toxin A/B uses antibody-coated colored particles to bind with target antigen in the diluted sample. The colored antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.
  • Clearly visible, easy-to-read results within 20 minutes
  • Less than one minute hands-on time
  • Integral control for added confidence
  • Excellent sensitivity and specificity
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 30°C
Description Xpect C. difficile Toxin A/B Test
Includes Test devices, conjugate reagent 1 (3.6mL), conjugate reagent 2 (3.0mL), specimen diluent, positive control (2.0mL), negative control (2.0mL), disposable transfer pipettes, dilution tubes, wooden applicator sticks
Quantity 20 Tests/Kit