Learn More
Description
Optimize your workflow by rapidly detecting Cryptosporidium-specific antigen from fresh, frozen, or preserved human fecal specimens using Thermo Scientific™ Xpect™ Cryptosporidium Test. Report accurate results in 15 minutes during all stages of infection—all from a single specimen.
- Clearly visible, easy-to-read results in just 15 minutes
- Less than one minute hands-on time
- 95.8 to 97.9% sensitivity
- Single and combination immunoassays available
- No specimen dilution required when using the recommended Formalin, SAF, or modified Cary-Blair w/Indicator transport media
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Xpect Cryptosporidium Test |
| Includes | Test devices, conjugate (2.5mL), specimen dilution buffer (2.5mL), procedure card, disposable transfer pipettes, dilution tubes |
| Quantity | 20 Tests/Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.