Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Xpect™ Flu A & B Test
GSA_VA

Catalog No. r24600
Change view
Click to view available options
Quantity:
20 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R24600 20 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R24600 Supplier Thermo Scientific™ Supplier No. R24600
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rapidly detect Influenza A and Influenza B viral antigens from human nasal wash, nasal swab and throat swab specimens, and get results in 15 minutes.

Rapidly detect Influenza A and Influenza B viral antigens from human nasal wash, nasal swab and throat swab specimens using Thermo Scientific™ Xpect™ Flu A & B Test. Maintain efficiency and productivity with a simple two-step procedure that offers accurate results in 15 minutes. When you need STAT testing or 24/7 coverage, choose Xpect.

Use this rapid, lateral flow immunoassay for the direct qualitative detection of Influenza A and Influenza B viral antigens. Reduce secondary testing, decrease unnecessary antibiotic use and guide appropriate antiviral therapy.


Accurate, Easy-to-Read Results
  • Improve patient management with accurate, easy-to-read results in just 15 minutes
  • Eliminate workflow bottlenecks with less than one minute hands-on time and a simple, walk-away procedure

Flexible Options
  • Flexible specimen storage, collection and transport options
  • Recent FDA clearance for analytical detection of cultured influenza A H7N9 virus

Xpect Flu A&B incorporates separate membrane strips for Influenza A and Influenza B. The patient specimen is diluted and added to both sample wells of the device. Antibody-coated color particles bind with target antigen in the diluted sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.

Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.

Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Xpect Flu A and B Test
Detectable Analytes Influenza
Sample Type Nasopharyngeal
Type Reagent
Includes Test devices, specimen diluent (20mL), dilution tubes, disposable transfer pipettes, flu A+/B- control swab and flu B+/A- control swab
Quantity 20 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.