Learn More
Thermo Scientific™ Xpect™ Giardia/Cryptosporidium Test
GSA_VA
Catalog No. 23313997 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 Tests/Kit
Catalog No. Quantity
23-313-997 20 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-313-997 Supplier Thermo Scientific™ Supplier No. R2450520
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rapidly detect Giardia- and Cryptosporidium-specific antigens from fresh, frozen, or preserved human fecal samples.
Optimize your workflow by rapidly detecting Giardia- and Cryptosporidium-specific antigens from fresh, frozen, or preserved human fecal specimens using Thermo Scientific™ Xpect™ Giardia/Cryptosporidium Test. Report accurate results in 15 minutes during all stages of infection—all from a single specimen.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Giardia- and Cryptosporidium-specific antigens. Enhance laboratory performance and efficiency with excellent clinical sensitivity and specificity and a simple walk-away procedure.
  • Clearly visible, easy-to-read results in just 15 minutes
  • Less than one minute hands-on time
  • 95.8 to 97.9% sensitivity
  • Single and combination immunoassays available
  • No specimen dilution required when using the recommended Formalin, SAF, or modified Cary-Blair w/Indicator transport media
Xpect Giardia/Cryptosporidium uses antibody-coated color particles to bind with target antigen in the diluted sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. A blue line will appear at the giardia test position if giardia antigen is present. If cryptosporidium antigen is present, a red line will appear at the cryptosporidium test position. These lines will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories.
Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description Xpect Giardia/Cryptosporidium Test
CE Marker Yes
Detectable Analytes Giardia and Cryptosporidium Antigen
Includes Test devices, conjugate (2.5mL), specimen dilution buffer (2.5mL), procedure card, disposable transfer pipettes, dilution tubes
Quantity 20 Tests/Kit