Learn More
Thermo Scientific™ Xpect™ Rotavirus Test
Catalog No. R24655 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 Tests/Kit
Catalog No. Quantity
R24655 20 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. R24655 Supplier Thermo Scientific™ Supplier No. R24655
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rapidly detect Rotavirus from human fecal samples.
Optimize your workflow by rapidly detecting rotavirus antigen in human fecal specimens using Thermo Scientific™ Xpect Rotavirus Test. Combine excellent clinical performance with a simple, two-step procedure that offers accurate results within 15 minutes.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Rotavirus antigen. Enhance laboratory performance and efficiency with excellent clinical sensitivity and specificity and a simple walk-away procedure.
  • Clearly visible, easy-to-read results in just 15 minutes
  • Less than one minute hands-on time
  • 100% sensitivity and specificity
  • Stock up with 18-month shelf life at room-temperature storage
Xpect Rotavirus uses antibody-coated color particles to bind with target antigen in the diluted sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Description Rotavirus Immunoassay
Quantity 20 Tests/Kit