Learn More
Zymo Research Corporation Xpedition™ Lysis/Stabilization Solution (40 ml)
SDP
Catalog No. 50444479
Encompass_Preferred
Xpedition Lysis/Stabilization Solution is used in conjunction with mechanical lysis for efficient extraction/preservation of DNA in sample lysates for long periods of time without the need for refrigeration.
Catalog No. 50-444-479 Supplier Zymo Research Corporation Supplier No. D6202140/DUP
Xpedition Lysis/Stabilization Solution is used in conjunction with mechanical lysis for efficient extraction/preservation of DNA in sample lysates for long periods of time without the need for refrigeration.