Learn More
Maine Molecular Quality Controls Inc Xpert BCR-ABL IS p210 Linearity Panel C207
SDP
Catalog No. NC3114538
Encompass
ABL1 endogenous control mRNA transcript when analyzed using the Xpert NPM1 Mutation assay on Cepheid GeneXpert Instrument Systems. The Xpert NPM1 Control Panel C194v1.1 consists of 5 components. Each component contains varying concentrations of NPM1 Type A, Type B or Type D mutant RNA transcript mixed with fixed concentrations of ABL1 and wildtype NPM1 transcripts to produce 5 levels of NPM1 Mutant RNA to ABL1 RNA; 0%, 1% NPM1 Type A, 1% NPM1 Type B, 1% NPM1 Type D, and 5% NPM1 Type A. The 0% level contains wildtype NPM1 and wildtype ABL1 transcripts only.
Catalog No. NC3114538 Supplier Maine Molecular Quality Controls Inc Supplier No. C207
ABL1 endogenous control mRNA transcript when analyzed using the Xpert NPM1 Mutation assay on Cepheid GeneXpert Instrument Systems. The Xpert NPM1 Control Panel C194v1.1 consists of 5 components. Each component contains varying concentrations of NPM1 Type A, Type B or Type D mutant RNA transcript mixed with fixed concentrations of ABL1 and wildtype NPM1 transcripts to produce 5 levels of NPM1 Mutant RNA to ABL1 RNA; 0%, 1% NPM1 Type A, 1% NPM1 Type B, 1% NPM1 Type D, and 5% NPM1 Type A. The 0% level contains wildtype NPM1 and wildtype ABL1 transcripts only.