Learn More
Maine Molecular Quality Controls Inc Xpert NPM1 Control Panel C194v1.1
SDP
Catalog No. NC3886298
Encompass
The Xpert NPM1 Control Panel C194v1.1 is intended for use as an assayed external quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the quantitative detection of the NPM1 Type A, Type B, and Type D mutant mRNA transcripts and
Catalog No. NC3886298 Supplier Maine Molecular Quality Controls Inc Supplier No. C194V1.1
The Xpert NPM1 Control Panel C194v1.1 is intended for use as an assayed external quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the quantitative detection of the NPM1 Type A, Type B, and Type D mutant mRNA transcripts and