Learn More
Maine Molecular Quality Controls Inc Xpert PML-RARA Control Panel C215
SDP
Catalog No. NC3754497
Encompass
The Xpert PML-RARA Control Panel C215 is an external reference material used to monitor the performance of in vitro laboratory nucleic acid testing procedures for the quantitative detection of PML-RARA transcript isoforms, bcr1, bcr2, bcr3, and ABL1 endogenous control mRNA transcript when analyzed using the Xpert PML-RARA assay on Cepheid GeneXpert Instrument Systems. The Xpert PML-RARA Control Panel C215 consists of 5 components. Each component contains varying concentrations of PML-RARA isoforms; bcr1, bcr2 or bcr3, mixed with a fixed concentration of ABL1 to produce five levels: PML-RARA 0%, PML-RARA bcr1 1%, PML-RARA bcr2 1%, PML-RARA bcr3 1%, and PML-RARA bcr1 20%. The 0% level contains only wildtype ABL1 transcript.
Catalog No. NC3754497 Supplier Maine Molecular Quality Controls Inc Supplier No. C215
The Xpert PML-RARA Control Panel C215 is an external reference material used to monitor the performance of in vitro laboratory nucleic acid testing procedures for the quantitative detection of PML-RARA transcript isoforms, bcr1, bcr2, bcr3, and ABL1 endogenous control mRNA transcript when analyzed using the Xpert PML-RARA assay on Cepheid GeneXpert Instrument Systems. The Xpert PML-RARA Control Panel C215 consists of 5 components. Each component contains varying concentrations of PML-RARA isoforms; bcr1, bcr2 or bcr3, mixed with a fixed concentration of ABL1 to produce five levels: PML-RARA 0%, PML-RARA bcr1 1%, PML-RARA bcr2 1%, PML-RARA bcr3 1%, and PML-RARA bcr1 20%. The 0% level contains only wildtype ABL1 transcript.