Learn More
Hach Company Xpert Xpress CoV-2/Flu/RSV
Catalog No. 5000206988 Shop All Hach Company Products
Encompass
Xpert Xpress CoV-2/Flu/RSV (RUO)-10 CartridgesSelf-contained single use cartridge for extraction amplification and detection of samples Includes FLU A1 2 B RSV SARS-COV-2 gene analysis For use with Cepheid GeneXpert analyzer
Catalog No. 50-002-06988 Supplier Hach Company Supplier No. LCE010
Xpert Xpress CoV-2/Flu/RSV (RUO)-10 CartridgesSelf-contained single use cartridge for extraction amplification and detection of samples Includes FLU A1 2 B RSV SARS-COV-2 gene analysis For use with Cepheid GeneXpert analyzer