Learn More
Hach Company Xpert Xpress SARS-CoV-2 Plus
Supplier:  Hach Company LCE009
Encompass
Xpert Xpress SARS-CoV-2 Plus-10 CartridgesSelf-contained single use cartridge for extraction amplification and detection of samples Includes SARS-COV-2 gene analysis For use with Cepheid GeneXpert analyzer
Catalog No. 50-002-07041