Learn More
Strem, An Ascensus Company CAS# 1310584-14-5. Chloro(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl) palladium(II), min. 98% [XPhos Palladacycle Gen. 2]. MFCD28100489. 1G
SDP
Catalog No. 501959499
Encompass
CAS# 1310584-14-5. 1G. Chloro(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl) palladium(II), min. 98% [XPhos Palladacycle Gen. 2]. MFCD28100489. Molecular Formula: C45H59ClNPPd. Molecular Weight: 786.80. Color/Form: off-white to beige pwdr. Strem# 46-0281. www.strem.com/catalog/v3/46-0281/51/
Catalog No. 50-195-9499 Supplier Strem, An Ascensus Company Supplier No. 4602811G
CAS# 1310584-14-5. 1G. Chloro(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl) palladium(II), min. 98% [XPhos Palladacycle Gen. 2]. MFCD28100489. Molecular Formula: C45H59ClNPPd. Molecular Weight: 786.80. Color/Form: off-white to beige pwdr. Strem# 46-0281. www.strem.com/catalog/v3/46-0281/51/