Learn More
Strem, An Ascensus Company CAS# 1445085-55-1. 5g. Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct, min. 98% [Xphos Palladacycle Gen. 3]. MFCD22666043
SDP
Catalog No. 502016702
Encompass
CAS# 1445085-55-1. 5g. Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct, min. 98% [Xphos Palladacycle Gen. 3]. MFCD22666043. Molecular Weight: 846.45. Molecular Formula: C46H62NO3PPdS. Color/form: white to off-white pwdr. Strem# 46-0320. http://www.strem.com/catalog/v/46-0320/
Catalog No. 50-201-6702 Supplier Strem, An Ascensus Company Supplier No. 4603205G
CAS# 1445085-55-1. 5g. Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct, min. 98% [Xphos Palladacycle Gen. 3]. MFCD22666043. Molecular Weight: 846.45. Molecular Formula: C46H62NO3PPdS. Color/form: white to off-white pwdr. Strem# 46-0320. http://www.strem.com/catalog/v/46-0320/