Learn More
Strem, An Ascensus Company CAS# 1599466-81-5. 250mg. Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [XPhos Palladacycle Gen. 4]. MFCD28411652
SDP
Catalog No. 502016732
Encompass
CAS# 1599466-81-5. 250mg. Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [XPhos Palladacycle Gen. 4]. MFCD28411652. Molecular Weight: 860.48. Molecular Formula: C47H64NO3PPdS. Color/form: white to off-white pwdr. Strem# 46-0327. http://www.strem.com/catalog/v/46-0327/
Catalog No. 50-201-6732 Supplier Strem, An Ascensus Company Supplier No. 460327250MG
CAS# 1599466-81-5. 250mg. Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [XPhos Palladacycle Gen. 4]. MFCD28411652. Molecular Weight: 860.48. Molecular Formula: C47H64NO3PPdS. Color/form: white to off-white pwdr. Strem# 46-0327. http://www.strem.com/catalog/v/46-0327/