Learn More
USA Scientific Inc Xplorer Electr 1 Chan Pipet
Catalog No. 502745086
Encompass
Eppendorf Xplorer Electronic Single Channel Pipette with charging adapter, 5-100 ul, Yellow Rocker
Catalog No. 50-274-5086 Supplier USA Scientific Inc Supplier No. 40610023
Eppendorf Xplorer Electronic Single Channel Pipette with charging adapter, 5-100 ul, Yellow Rocker