Learn More
USA Scientific Inc Xplorer Electr 12ch , 5-100 uL
Catalog No. 502745075
Encompass
Eppendorf Xplorer Electronic 12-Channel Pipette with charging adapter, 5-100 ul, Yellow Rocker
Catalog No. 50-274-5075 Supplier USA Scientific Inc Supplier No. 40610139
Eppendorf Xplorer Electronic 12-Channel Pipette with charging adapter, 5-100 ul, Yellow Rocker