Learn More
Molecular Devices Molecular Devices Reporter Assay Kit, Homogeneous Glow, 5 fg
Catalog No. 5000014639
Encompass_Preferred
The SpectraMax Glo Steady-Luc Reporter Assay Kit is a homogeneous, glow-type luminescent assay for quantifying firefly luciferase expression in mammalian cells. It delivers a stable, high-intensity signal optimized for microplate readers and is suitable for reporter gene assays, gene expression studies, and signal transduction research.
  • Homogeneous glow assay format compatible with microplate readers.
  • Produces a stable, long-lasting luminescent signal for extended read times.
  • High sensitivity down to approximately 5 fg of luciferase per well.
  • Wide linear dynamic range spanning about seven orders of magnitude.
  • Reagent volume sufficient for approximately ten 96-well plate assays.
  • Optimized for use with microplate reader protocols and preconfigured software routines.
Catalog No. 50-000-14639 Supplier Molecular Devices Supplier No. R8353
The SpectraMax Glo Steady-Luc Reporter Assay Kit is a homogeneous, glow-type luminescent assay for quantifying firefly luciferase expression in mammalian cells. It delivers a stable, high-intensity signal optimized for microplate readers and is suitable for reporter gene assays, gene expression studies, and signal transduction research.
  • Homogeneous glow assay format compatible with microplate readers.
  • Produces a stable, long-lasting luminescent signal for extended read times.
  • High sensitivity down to approximately 5 fg of luciferase per well.
  • Wide linear dynamic range spanning about seven orders of magnitude.
  • Reagent volume sufficient for approximately ten 96-well plate assays.
  • Optimized for use with microplate reader protocols and preconfigured software routines.