Learn More
Aviva Systems Biology Corp Xpnpep2 Antibody - C-terminal region (ARP41526_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41526_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Xpnpep2. It was validated on Western Blot. . Alternate names: 9030008G12Rik, mAPPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-939