Learn More
Aviva Systems Biology Corp XPNPEP3 Antibody - N-terminal region (ARP57593_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57593_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XPNPEP3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human XPNPEP3. Alternate names: APP3, NPHPL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-338