Learn More
Aviva Systems Biology Corp XPO1 Antibody - middle region (ARP40466_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40466_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XPO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human XPO1. Alternate names: CRM1, DKFZp686B1823, emb, exp1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57468