Learn More
Waters Corp Waters Sep-Pak XPoSure Plus Short Cartridge, 350 mg Sorbent, DNPH-coated Silica, 20/pk
Catalog No. 50785643 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is designed for monitoring aldehydes in indoor air by trapping them with acidified 2,4-dinitrophenylhydrazine (DNPH) to form stable hydrazone derivatives. These derivatives are then eluted and analyzed using HPLC. It features high-quality packings for sample preparation.
  • Quantifies formaldehyde (0.001 to 5 ppmv) in workplace and indoor air.
  • High-quality packings for optimal surface area, pore size, and particle distribution.
  • Sealed in non-contaminating poly-foil pouches to maintain integrity.
  • Suitable for manual and robotic applications.
  • Contains acidified 2,4-dinitrophenylhydrazine (DNPH)-coated silica.
  • Designed for flow rates up to 1.5 L/min with typical personal pumps.
  • Collection efficiency: >95% for sampling rates up to 1.5 L/min.
  • Sampling temperature: 10°C to 100°C.
Catalog No. 50-785-643 Supplier Waters Corp Supplier No. WAT047205
This product is designed for monitoring aldehydes in indoor air by trapping them with acidified 2,4-dinitrophenylhydrazine (DNPH) to form stable hydrazone derivatives. These derivatives are then eluted and analyzed using HPLC. It features high-quality packings for sample preparation.
  • Quantifies formaldehyde (0.001 to 5 ppmv) in workplace and indoor air.
  • High-quality packings for optimal surface area, pore size, and particle distribution.
  • Sealed in non-contaminating poly-foil pouches to maintain integrity.
  • Suitable for manual and robotic applications.
  • Contains acidified 2,4-dinitrophenylhydrazine (DNPH)-coated silica.
  • Designed for flow rates up to 1.5 L/min with typical personal pumps.
  • Collection efficiency: >95% for sampling rates up to 1.5 L/min.
  • Sampling temperature: 10°C to 100°C.