Learn More
OPTIMUM PROCESSING INC XpressVent bag 10L
Catalog No. NC1797512
Encompass
This item has a minimum order qty of 10 per supplier requirements.
XpressVent containers are pre-sterilized disposable assemblies designed to ease liquid handling for various additions, storage and processing steps. They are less expensive than rigid bottles and are collapsible, They are available with a strong hanging or carry handle and in a clear or opaque brown light restricting LDPE material. This is a new level of convenience to replace glass, rigid PETG, PE or PP bottles and SS stopper assemblies commonly found in laboratories, pilot plant, development and research labs.
Catalog No. NC1797512 Supplier OPTIMUM PROCESSING INC Supplier No. 40170
This item has a minimum order qty of 10 per supplier requirements.
XpressVent containers are pre-sterilized disposable assemblies designed to ease liquid handling for various additions, storage and processing steps. They are less expensive than rigid bottles and are collapsible, They are available with a strong hanging or carry handle and in a clear or opaque brown light restricting LDPE material. This is a new level of convenience to replace glass, rigid PETG, PE or PP bottles and SS stopper assemblies commonly found in laboratories, pilot plant, development and research labs.