Learn More
CEM Corporation XPRESS VENT PLUG
SDP
Catalog No. NC9272560
Encompass
MARS Xpress Vessel Vent Plug. This TFM Vent plug makes the seal between the liner and the cover. Used with MARS Xpress vessels.
Catalog No. NC9272560 Supplier CEM Corporation Supplier No. 212020
MARS Xpress Vessel Vent Plug. This TFM Vent plug makes the seal between the liner and the cover. Used with MARS Xpress vessels.