Learn More
XpressBio XpressMyco PCR Plus Detection Kit, 100 Reactions
SDP
Supplier:  XpressBio XMYCG100
Encompass_Preferred
The XpressMyco PCR Plus Detection Kit offers high specificity and sensitivity to minimize false positives while ensuring coverage over 200 strains of Mycoplasmas for quick and reliable routine screening of cell cultures less than 1 hour.
Catalog No. NC2956717