Learn More
TARGETMOL CHEMICALS INC XR-11576 HCL 5MG
SDP
Catalog No. 502580705
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. XR-11576 HCl is a novel orally active dual topoisomerase I and II inhibitor with antitumor activity.
Catalog No. 50-258-0705 Supplier TARGETMOL CHEMICALS INC Supplier No. T26339L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. XR-11576 HCl is a novel orally active dual topoisomerase I and II inhibitor with antitumor activity.