Learn More
Medchemexpress LLC XR8-89 | 2817811-16-6 | C29H36N4O2S | 1 ML
SDP
Catalog No. 5000375852
Encompass_Preferred
XR8-89 is a potent papain-like protease (PLpro) inhibitor with an IC50 value of 0.1 μM. It induces conformational changes in SARS-CoV-2 papain-like protease, inhibiting SARS-CoV-2 replication and can be used for SARS-CoV-2 research. It is soluble in DMSO at 50 mg/mL.
  • Potent papain-like protease (PLpro) inhibitor
  • Induces conformational changes in SARS-CoV-2 papain-like protease
  • Inhibits SARS-CoV-2 replication
  • Suitable for SARS-CoV-2 research
  • Soluble in DMSO
Catalog No. 50-003-75852 Supplier Medchemexpress LLC Supplier No. HY15078410MM/1ML
XR8-89 is a potent papain-like protease (PLpro) inhibitor with an IC50 value of 0.1 μM. It induces conformational changes in SARS-CoV-2 papain-like protease, inhibiting SARS-CoV-2 replication and can be used for SARS-CoV-2 research. It is soluble in DMSO at 50 mg/mL.
  • Potent papain-like protease (PLpro) inhibitor
  • Induces conformational changes in SARS-CoV-2 papain-like protease
  • Inhibits SARS-CoV-2 replication
  • Suitable for SARS-CoV-2 research
  • Soluble in DMSO