Learn More
Premier Lab Supply 40mm XRF Sample Cups – PN# SC-4140
SDP
Catalog No. NC0449802
Encompass_Preferred
40mm Single-Open Ended XRF Sample Cup with Cup & 2 Rings (PN# SC-4140) – Suitable for Universal – OD: 40mm; ID: 32mm; Height: 23mm; Volume: 18mL – 100 per bag
Catalog No. NC0449802 Supplier Premier Lab Supply Supplier No. SC4140
40mm Single-Open Ended XRF Sample Cup with Cup & 2 Rings (PN# SC-4140) – Suitable for Universal – OD: 40mm; ID: 32mm; Height: 23mm; Volume: 18mL – 100 per bag