Learn More
Med Vet International McKesson Sterile X-ray Detectable Gauze Sponge, 16-Ply, 4 x 8 Inch, 16-42447, 800/Case
Catalog No. 503043865
Encompass_Preferred
X-Ray Detectable Gauze Sponge McKesson Cotton 16-Ply 4 X 8 Inch Rectangle Sterile Radiopaque design contains blue X-ray-detectable barium sulfate strands for easy identification Made from 16-ply 100% cotton Sterile sponge is intended for single-use applications 4 x 8 inches
Catalog No. 50-304-3865 Supplier Med Vet International Supplier No. 944343CS
X-Ray Detectable Gauze Sponge McKesson Cotton 16-Ply 4 X 8 Inch Rectangle Sterile Radiopaque design contains blue X-ray-detectable barium sulfate strands for easy identification Made from 16-ply 100% cotton Sterile sponge is intended for single-use applications 4 x 8 inches