Learn More
Enzo Life Sciences XRCC1 (human) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210539R100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human XRCC1 (X-ray repair cross-complementing group 1) fused to a His-tag. Species reactivity: Human. Applications: ICC, IP, WB. Formulation: Liquid. Antiserum containing 0.1% sodium azide.
Catalog No. 50-200-8130