Learn More
Aviva Systems Biology Corp XRCC2 Antibody - middle region (ARP58558_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58558_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XRCC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human XRCC2. Alternate names: DKFZp781P0919ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62354