Learn More
Aviva Systems Biology Corp XRCC3 Antibody - middle region (ARP61275_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61275_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XRCC3. It was validated on Western Blot. . Alternate names: CMM6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse
Catalog No. 50-287-322