Learn More
Premier Lab Supply 45mm XRF Sample Cups – PN# SC-4345
SDP
Catalog No. NC0588941
Encompass_Preferred
45mm Double-Open Ended XRF Sample Cup with Cup, Ring & Cap (PN# SC-4345) – Suitable for Philips/Panalytical – OD: 45mm; ID: 38mm; Height: 34mm; Volume: 36mL – 100 per bag
Catalog No. NC0588941 Supplier Premier Lab Supply Supplier No. SC4345
45mm Double-Open Ended XRF Sample Cup with Cup, Ring & Cap (PN# SC-4345) – Suitable for Philips/Panalytical – OD: 45mm; ID: 38mm; Height: 34mm; Volume: 36mL – 100 per bag