Learn More
Premier Lab Supply 47mm XRF Sample Cups – PN# SC-8047
SDP
Catalog No. NC0684824
Encompass_Preferred
47mm Single-Open Ended XRF Sample Cup with Cup & Ring (PN# SC-8047) – Suitable for Horiba; XOS – OD: 47mm; ID: 36mm; Height: 19mm; Volume: 15mL – 100 per bag
Catalog No. NC0684824 Supplier Premier Lab Supply Supplier No. SC8047
47mm Single-Open Ended XRF Sample Cup with Cup & Ring (PN# SC-8047) – Suitable for Horiba; XOS – OD: 47mm; ID: 36mm; Height: 19mm; Volume: 15mL – 100 per bag