Learn More
Premier Lab Supply 32mm XRF Sample Cups – PN# SC-7332
SDP
Catalog No. NC0930430
Encompass_Preferred
32mm Double-Open Ended XRF Sample Cup with Inner Cup, Cap & Aluminum Holder (PN# SC-7332) – Suitable for Oxford LABX – OD: 32mm; ID: 30mm; Height: 38mm; Volume: 18mL – 100 per bag
Catalog No. NC0930430 Supplier Premier Lab Supply Supplier No. SC7332
32mm Double-Open Ended XRF Sample Cup with Inner Cup, Cap & Aluminum Holder (PN# SC-7332) – Suitable for Oxford LABX – OD: 32mm; ID: 30mm; Height: 38mm; Volume: 18mL – 100 per bag